Learn More
Sino Biological Zaire ebolavirus (Guinea 2014) VP40 Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 101299T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Zaire ebolavirus (Guinea 2014) VP40, and purified by antigen affinity chromatography.
Catalog No. 50-172-1598