Learn More
Sino Biological Recombinant Human FABP2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10130H08E100
Encompass_Preferred
A DNA sequence encoding the human FABP2 (P12104-1) (Met 1-Asp 132) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4096