Learn More
Sino Biological Anti-pIgR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10131RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PIGR extracellular domain (rh PIGR; Catalog#10131-H08H; NP_002635.2; Met1-Arg638,365Ser/Gly). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-1634