Learn More
Sino Biological Recombinant Anti-Meprin alpha/MEP1A Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10133R009100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human MEP1A (rh MEP1A; Catalog#10133-H08H; NP_005579.2; Met1-Gln601).
Catalog No. 50-172-1655