Learn More
Sino Biological Anti-MCP-1/CCL2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10134MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MCP-1 / CCL2 (rh MCP-1 / CCL2; Catalog#10134-H08Y; NP_002973.1; Gln24-Thr99). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1689