Learn More
Sino Biological Anti-MCP-1/CCL2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10134T26100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MCP-1/CCL2 (rh MCP-1/CCL2; Catalog#10134-H08Y; NP_002973.1; Gln24-Thr99). MCP-1/CCL2 specific IgG was purified by Human MCP-1/CCL2 affinity chromatography.
Catalog No. 50-172-1710