Learn More
Sino Biological Anti-Lambda 5/IGLL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101342T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human IGLL1/CD179b, and purified by antigen affinity chromatography.
Catalog No. 50-172-1668