Learn More
Sino Biological Anti-MEK1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101351T38100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MEK1/MAP2K1/MKK1, and purified by antigen affinity chromatography.
Catalog No. 50-172-1716