Learn More
Sino Biological Recombinant Human VEGFR1/FLT1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10136H08H1100
Encompass_Preferred
A DNA sequence encoding the human FLT1 (NP_002010.2) (Met1-Ile328) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4107