Learn More
Sino Biological Anti-BVES Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101360T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human BVES / POPDC1, and purified by antigen affinity chromatography.
Catalog No. 50-172-1722