Learn More
Sino Biological Anti-IL-1 beta Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10139RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL1B (rh IL1B; Catalog#10139-HNAE; NP_000567.1; Ala 117-Ser 269). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-1794