Learn More
Sino Biological Recombinant Human L1CAM Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10140H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD171 (NP_000416.1) (Met 1-Glu 1120) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4117