Learn More
Sino Biological Anti-L1CAM Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10140MM03A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD171 / N-CAML1 / L1CAM (rh CD171 / N-CAML1 / L1CAM; Catalog#10140-H08H; NP_000416.1; Met1-Glu1120) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-1813