Learn More
Sino Biological Recombinant Anti-L1CAM Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10140R014F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD171 / N-CAML1 / L1CAM (rh CD171 / N-CAML1 / L1CAM; Catalog#10140-H08H; NP_000416.1; Met 1-Glu 1120) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-1824