Learn More
Sino Biological Anti-Antithrombin III Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10142RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Serpin C1 / SERPINC1 (rh Serpin C1; Catalog#10142-H08H; NP_000479.1; Met 1-Lys 464). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-1848