Learn More
Sino Biological Recombinant Human CHL-1/L1CAM-2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10143H08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CHL1 (AAI04919.1) (Met 1-Gln 1080) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4122