Learn More
Sino Biological Anti-5T4/TPBG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101430T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human TPBG, and purified by antigen affinity chromatography.
Catalog No. 50-172-1857