Learn More
Sino Biological Recombinant Anti-Cadherin 8 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10144R032100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cadherin-8 / CDH8 (rh Cadherin-8 / CDH8; Catalog#10144-H08H; NP_001787.2; Met1-Met621).
Catalog No. 50-172-1880