Learn More
Sino Biological Recombinant Human TLR4 Protein (ECD, His Tag)
Supplier:  Sino Biological 10146H08B100
Encompass_Preferred
A DNA sequence encoding the human TLR4 (Met1-Lys631) (O00206-1) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4126