Learn More
Sino Biological Anti-CD31/PECAM-1 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10148MM02F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD31 / PECAM1 (rhCD31; Catalog#10148-H08H; Met 1-Lys 601; EAW94208.1) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-1934