Learn More
Sino Biological Recombinant Human Endoglin/CD105 Protein (ECD, His Tag)
Supplier:  Sino Biological 10149H08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD105 (NP_001108225.1) (Met 1-Gly 586) was expressed, with C-terminal fused polyhistidine tag.
Catalog No. 50-161-4140