Learn More
Sino Biological Anti-Endoglin/CD105 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10149MM13A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Endoglin / CD105 (rh Endoglin / CD105 ; Catalog#10149-H08H; NP_001108225.1; Met1-Gly586) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-2003