Learn More
Sino Biological Recombinant Anti-Endoglin/CD105 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10149R103A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Endoglin / CD105 (rh Endoglin / CD105; Catalog#10149-H08H; NP_001108225.1; Met1-Gly586) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-2014