Learn More
Sino Biological Recombinant Anti-Endoglin/CD105 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10149R103F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Endoglin / CD105 (rh CD105; Catalog#10149-H08H; Met 1-Gly 586; NP_001108225.1) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-2018