Learn More
Sino Biological Anti-Endoglin/CD105 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10149T26100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Endoglin/CD105 (rh Endoglin/CD105 ; Catalog#10149-H08H; NP_001108225.1; Met1-Gly586). Endoglin/CD105 specific IgG was purified by Human Endoglin/CD105 affinity chromatography.
Catalog No. 50-172-2028