Learn More
Sino Biological Anti-Cadherin-6/K-cadherin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10150T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Cadherin-6 / CDH6 (rh Cadherin-6 / CDH6; Catalog#10150-H08H; NP_004923.1; Met1-Ala615). Cadherin-6 / CDH6 specific IgG was purified by Human Cadherin-6 / CDH6 affinity chromatography.
Catalog No. 50-172-2046