Learn More
Sino Biological Anti-CD40/TNFRSF5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101510T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CD40/TNFRSF5, and purified by antigen affinity chromatography.
Catalog No. 50-172-2049