Learn More
Sino Biological Anti-KMT1B/SUV39H2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101528T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SUV39H2, and purified by antigen affinity chromatography.
Catalog No. 50-172-2070