Learn More
Sino Biological Recombinant Human Complement C2 Protein (hFc Tag)
Supplier:  Sino Biological 10154H02H20
Encompass_Preferred
A DNA sequence encoding the human complement component 2 (C2) precursor (NP_000054.2) (Met 1-Leu 752) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4144