Learn More
Sino Biological Recombinant Anti-Complement C2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10154R002100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human complement component 2 (rh C2; Catalog#10154-H08H; Met 1-Leu 752; NP_000054.2).
Catalog No. 50-172-2092