Learn More
Sino Biological Recombinant Human IL-37 Protein, HPLC-verified
Supplier:  Sino Biological 10155HNAE20
Encompass_Preferred
A DNA sequence encoding the mature form of human IL1F7 isoform A (Q9NZH6-2) (Lys 27-Asp 192) was expressed and purified, with an initial Met.
Catalog No. 50-161-4147