Learn More
Sino Biological Anti-IL-37 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10155MM20100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL37 / IL1F7 / IL-1H4 (rh IL37 / IL1F7 / IL-1H4; Catalog#10155-HNAE; Q9NZH6-2; Lys27-Asp192). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2106