Learn More
Sino Biological Anti-Epsin 3/EPN3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101551T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human EPN3, and purified by antigen affinity chromatography.
Catalog No. 50-172-2097