Learn More
Sino Biological Recombinant Human IL1RAPL2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10156H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Glu 356) of human IL1R9 (NP_059112.1) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-4151