Learn More
Sino Biological Anti-IL1RAPL2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10156MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-1R9 / IL1RAPL2 (rh IL-1R9 / IL1RAPL2; Catalog#10156-H08H; NP_059112.1; Met1-Glu356). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.
Catalog No. 50-172-2127