Learn More
Sino Biological Anti-CDC37 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101589T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CDC37, and purified by antigen affinity chromatography.
Catalog No. 50-172-2151