Learn More
Sino Biological Anti-Cathepsin Z/CTSZ Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10159T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CTSZ / CTSX / Cathepsin Z (rh CTSZ / CTSX / Cathepsin Z; Catalog#10159-H08H; Q9UBR2; Met1-Val303). CTSZ / CTSX / Cathepsin Z specific IgG was purified by Human CTSZ / CTSX / Cathepsin Z affinity chromatography.
Catalog No. 50-172-2166