Learn More
Sino Biological Anti-CDK3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101592T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CDK3, and purified by antigen affinity chromatography.
Catalog No. 50-172-2157