Learn More
Sino Biological Anti-Factor D Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10160T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FACTOR D/Adipsin (rh FACTOR D/Adipsin; Catalog#10160-H08H; P00746; Ile26-Ala253). FACTOR D/Adipsin specific IgG was purified by Human FACTOR D/Adipsin affinity chromatography.
Catalog No. 50-172-2189