Learn More
Sino Biological Recombinant Human M-CSFR/CSF1R Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10161H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Glu512) of human CSF1R (NP_005202.2) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4165