Learn More
Sino Biological Recombinant Human M-CSFR/CSF1R Protein (Domain I&II&III, His Tag)
Supplier:  Sino Biological 10161H08H1200
Encompass_Preferred
A DNA sequence encoding the human CSF1R (NP_005202.2) (Met1-Ser290) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4167