Learn More
Sino Biological Recombinant Human M-CSFR/CSF1R Protein
Supplier:  Sino Biological 10161HCCH200
Encompass_Preferred
The mature form of human M-CSFR (NP_005202.2) extracellular domain (Met 1-Glu 512) with quinary-aa peptide (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-161-4176