Learn More
Sino Biological Recombinant Human M-CSFR/CSF1R Protein, HPLC-verified
Supplier:  Sino Biological 10161HNAH100
Encompass_Preferred
A DNA sequence encoding the human CSF1R (NP_005202.2) (Met1-Glu512) was expressed.
Catalog No. 50-161-4177