Learn More
Sino Biological Anti-M-CSFR/CSF1R Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10161MM12100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human M-CSFR extracellular domain (rh M-CSFR; Catalog#10161-H08H; Met 1-Glu 512; NP_005202.2). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2206