Learn More
Sino Biological Anti-M-CSFR/CSF1R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10161RP03100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MCSFR extracellular domain (rh MCSFR; Catalog#10161-H08H; Met 1-Glu 512; NP_005202.2) MCSFR specific IgG was purified by human MCSFR affinity chromatography.
Catalog No. 50-172-2219