Learn More
Sino Biological Recombinant Human MDC/CCL22 Protein (His Tag)
Supplier:  Sino Biological 10163H07E50
Encompass_Preferred
A DNA sequence encoding the secreeted form of human CCL22 (O00626) (Gly 25-Gln 93) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-4180