Learn More
Sino Biological Recombinant Human IGF1R Protein (ECD, His Tag)
Supplier:  Sino Biological 10164H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Asn932) of human IGF1R (NP_000866.1) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4181