Learn More
Sino Biological Recombinant Human IGF1R Protein (His & GST Tag)
Supplier:  Sino Biological 10164H20B150
Encompass_Preferred
A DNA sequence encoding the human IGF1R (NP_000866.1) cytoplasmic domain (Met 954-Cys 1367) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-4184