Learn More
Sino Biological Anti-IGF1R Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10164MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IGF1R / CD221 extracellular domain (rhIGF1R; Catalog#10164-H08H; Met 1-Asn 932; NP_000866.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2243