Learn More
Sino Biological Recombinant Anti-IGF1R Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10164R011100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IGF1R / CD221 extracellular domain (rh IGF1R; Catalog#10164-H08H; Met 1-Asn 932; NP_000866.1).
Catalog No. 50-172-2250